Merck
  • Double-Track Electrochemical Green Approach for Simultaneous Dissolution Profiling of Naproxen Sodium and Diphenhydramine Hydrochloride.
Double-Track Electrochemical Green Approach for Simultaneous Dissolution Profiling of Naproxen Sodium and Diphenhydramine Hydrochloride.
Journal of pharmaceutical and biomedical analysis (2017-09-08)
Mostafa A Shehata, Esraa M Fawaz, Mohamed K Abd El-Rahman, Ezzat M Abdel-Moety
ABSTRACT
Acquisition of the dissolution profiles of more than single active ingredient in a multi-analyte pharmaceutical formulation is a mandatory manufacturing practice that is dominated by utilization of the off-line separation-based chromatographic methods. This contribution adopts a new "Double-Track" approach with the ultimate goal of advancing the in-line potentiometric sensors to their most effective applicability for simultaneous acquisition of the dissolution profiles of two active ingredients in a binary pharmaceutical formulation. The unique abilities of these sensors for real-time measurements is the key driver for adoption of "green analytical chemistry" (GAC) principles aiming to expand the application of eco-friendly analytical methods With the aim of performing a side-by-side comparison, this work investigates the degree of adherence of ISEs to the 12 principles of GAC in multicomponent dissolution profiling with respect to the HPLC. For the proof of concept, a binary mixture of naproxen sodium (NAPR) and diphenhydramine hydrochloride (DIPH) marketed as Aleve pm
Sigma-Aldrich
Calix[6]arene, 97%