Merck
  • Overexpression of the transmembrane protein BST-2 induces Akt and Erk phosphorylation in bladder cancer.
Overexpression of the transmembrane protein BST-2 induces Akt and Erk phosphorylation in bladder cancer.
Oncology letters (2017-07-12)
Yoshinori Shigematsu, Naohide Oue, Yuri Nishioka, Naoya Sakamoto, Kazuhiro Sentani, Yohei Sekino, Shoichiro Mukai, Jun Teishima, Akio Matsubara, Wataru Yasui
ABSTRACT
Bladder cancer, the majority of which is urothelial carcinoma (UC), is a common malignancy worldwide. Genes encoding transmembrane/secretory proteins expressed specifically in certain cancers may be ideal biomarkers for cancer diagnosis and may represent therapeutic targets. In the present study, the expression and function of the bone marrow stromal cell antigen 2
Sigma-Aldrich
Sample Buffer, Laemmli 2× Concentrate
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-BST2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution