Merck
  • Effect of Heteroatom Substitution on Transport in Alkanedithiol-Based Molecular Tunnel Junctions: Evidence for Universal Behavior.
Effect of Heteroatom Substitution on Transport in Alkanedithiol-Based Molecular Tunnel Junctions: Evidence for Universal Behavior.
ACS nano (2016-12-10)
Zuoti Xie, Ioan Bâldea, Stuart Oram, Christopher E Smith, C Daniel Frisbie
ABSTRACT
The transport properties of molecular junctions based on alkanedithiols with three different methylene chain lengths were compared with junctions based on similar chains wherein every third -CH
Sigma-Aldrich
2,2′-Thiodiethanethiol, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
1,11-Undecanedithiol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
1,5-Pentanedithiol, 96%
Pricing and availability is not currently available.