Merck
  • Native umbilical cord matrix stem cells express hepatic markers and differentiate into hepatocyte-like cells.
Native umbilical cord matrix stem cells express hepatic markers and differentiate into hepatocyte-like cells.
Gastroenterology (2008-02-05)
David Campard, Philippe A Lysy, Mustapha Najimi, Etienne Marc Sokal
ABSTRACT
Umbilical cord matrix stem cells (UCMSCs) are able to differentiate into mesodermal and ectodermal lineages. The present study investigates the differentiation potential of human UCMSCs into hepatic lineage. We isolated human UCMSCs and characterized them in vitro by measuring their expansion potential, by assessing expression of mesenchymal stem cell (MSC) markers, and by evaluating their ability to differentiate into adipocytes and osteocytes. UCMSCs were thereafter subjected to a hepatogenic differentiation protocol. Expression of hepatic and MSC markers in differentiated cells was analyzed by reverse-transcription polymerase chain reaction, flow cytometry, and immunocytochemical assays and compared with undifferentiated UCMSCs and freshly isolated liver cells. UCMSCs were transplanted into livers of hepatectomized-SCID mice, and engraftment capacity was investigated by detection of human nucleus and mitochondria and human hepatic-specific proteins. In vitro expanded UCMSCs constitutively expressed markers of hepatic lineage, including albumin, alpha-fetoprotein, cytokeratin-19, connexin-32, and dipeptidyl peptidase IV. In vitro-differentiated UCMSCs exhibited hepatocyte-like morphology, up-regulated several hepatic markers, stored glycogen, produced urea, and exhibited an inducible CYP 3A4 activity. However, absence of some hepatic markers in differentiated UCMSCs, as HepPar1 or hepatocyte nuclear factor 4 (HNF-4), implied that their differentiation did not reach the level of mature hepatocytes. We also noticed that differentiated UCMSCs partially preserved MSC markers. Engraftment capacity of UCMSCs was observed, and expression of human albumin and alpha-fetoprotein was detected 2, 4, and 6 weeks after transplantation in mice livers, while cytokeratin 19 was completely down-regulated. We conclude that UCMSCs, with a newly demonstrated endodermic differentiation potential, might be an alternative source for liver-directed cell therapies.
Sigma-Aldrich
Anti-Connexin 32 Antibody, clone M12.13, clone M12.13, Chemicon®, from mouse
Sigma-Aldrich
Anti-Cytokeratin 7 Antibody, clone RCK105, clone RCK105, Chemicon®, from mouse