Merck
  • Olanzapine for treatment and prevention of acute chemotherapy-induced vomiting in children: a retrospective, multi-center review.
Olanzapine for treatment and prevention of acute chemotherapy-induced vomiting in children: a retrospective, multi-center review.
Pediatric blood & cancer (2014-10-21)
Jacqueline Flank, Jennifer Thackray, Danelle Nielson, Amanda August, Tal Schechter, Sarah Alexander, Lillian Sung, L Lee Dupuis
ABSTRACT
This retrospective review provides preliminary data regarding the safety and efficacy of olanzapine for chemotherapy-induced vomiting (CIV) control in children. Children <18 years old who received olanzapine for acute chemotherapy-induced nausea and vomiting (CINV) control from December 2010 to August 2013 at four institutions were identified. Patient characteristics, chemotherapy, antiemetic prophylaxis, olanzapine dosing, CIV control, liver function test results and adverse events were abstracted from the health record. Toxicity was graded using CTCAEv4.03. Sixty children (median age 13.2 years; range: 3.10-17.96) received olanzapine during 158 chemotherapy blocks. Olanzapine was most often (59%) initiated due to a history of poorly controlled CINV. The mean initial olanzapine dose was 0.1 mg/kg/dose (range: 0.026-0.256). Most children who received olanzapine beginning on the first day of the chemotherapy block experienced complete CIV control throughout the acute phase (83/128; 65%). There was no association between the olanzapine dose/kg and complete CIV control (OR 1.01; 95% CI: 0.999-1.020; P = 0.091). Sedation was reported in 7% of chemotherapy blocks and was significantly associated with increasing olanzapine dose (OR: 1.17; 95% CI: 1.08-1.27; P = 0.0001). Of the 25 chemotherapy blocks where ALT and/or AST were reported more than once, grade 1-3 elevations were observed in five. The mean weight change in 31 children who received olanzapine during more than one chemotherapy block was 0% (range: -22 to +18). Olanzapine may be an important option to improve CIV control in children. Prospective controlled evaluation of olanzapine for CINV prophylaxis in children is warranted.
Product Description
USP
Olanzapine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)
Olanzapine, European Pharmacopoeia (EP) Reference Standard
Olanzapine for system suitability, European Pharmacopoeia (EP) Reference Standard