Merck
  • Immunohistochemical and immunochemical characterization of a new endothelial cell-specific antigen.
Immunohistochemical and immunochemical characterization of a new endothelial cell-specific antigen.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1986-02-01)
J U Alles, K Bosslet
ABSTRACT
A new monoclonal antibody (moab BW 200) of IgG3 kappa-isotype was generated which recognizes an epitope located on an antigen molecule restricted to human neoplastic and non-neoplastic endothelial cells. The molecular weight of the antigen was determined using immunoprecipitation analysis followed by SDS-PAGE. Despite its similar molecular weight to FVIII-RAG, the antigen detected by moab BW 200 was shown to be different from FVIII-RAG.
Sigma-Aldrich
CD31 (EP78) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
CD31 (JC70) Mouse Monoclonal Antibody