Merck
  • [Glycoconjugates on the basis of polyacrylamide--tools for studying lectins, antibodies, and glycosyltransferases in glycobiology, cytochemistry, and histochemistry].
[Glycoconjugates on the basis of polyacrylamide--tools for studying lectins, antibodies, and glycosyltransferases in glycobiology, cytochemistry, and histochemistry].
Bioorganicheskaia khimiia (1996-09-01)
N V Bovin
ABSTRACT
The synthesis, physicochemical characteristics, and application for studying the carbohydrate-binding molecules of some analogues of human cell glycoconjugates, neoglycoconjugates, were reviewed. An approach to the synthesis of the polyacrylamide derivatives of carbohydrates based on the interaction of fully activated polyacrylic acid with omega-aminoalkyl glycosides was described. It provides highly reproducible results, is more simple than the previously known methods of the synthesis of such derivatives, and expands the range of synthetic possibilities because it can provide both the sugar-polymer type molecules and conjugates bearing various labels and effectors, sorbents, glycosurfaces, etc. In the first part of the review, the synthesis of polyacrylamide conjugates and their physicochemical properties were described. In its second part, the synthesis of some complex constructions, such as pseudoglycoproteins, pseudomucins, glycoparticles, and glycosurfaces, was outlined. Some examples of the application of the described conjugates in various fields of glycobiology were simultaneously discussed. Prospects of the further development of the presented approach in glycotechnology and medicine were also described.
Sigma-Aldrich
Polyacrylamide, average Mn 40,000
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Polyacrylamide, average Mn 150,000