Merck
  • A transition-state-analog inhibitor influences zinc-binding by Aeromonas aminopeptidase.
A transition-state-analog inhibitor influences zinc-binding by Aeromonas aminopeptidase.
Biochemical and biophysical research communications (1985-08-15)
J O Baker, J M Prescott
ABSTRACT
The transition-state-analog inhibitor, 1-butaneboronic acid, markedly enhances the uptake of one g-atom of Zn2+ ions from a metal ion buffer system by Zn-depleted Aeromonas aminopeptidase. In contrast, a substrate-analog inhibitor, n-valeramide, does not perturb the equilibrium between Zn2+ ions and the enzyme in a metal ion buffer system. These results establish a role for metal ions in the binding of 1-butaneboronic acid to Aeromonas amino-peptidase and strongly imply that a bound Zn2+ ion interacts directly with substrate during catalysis but not during initial binding of substrate.
Product Description
Supelco
Butylboronic acid, for GC derivatization, LiChropur, ≥96.0% (T)
Sigma-Aldrich
Butylboronic acid, 97%