Merck
  • Characterization of adsorption on the stationary phase using high-performance immunoaffinity chromatography.
Characterization of adsorption on the stationary phase using high-performance immunoaffinity chromatography.
Journal of chromatography (1987-12-25)
R G Nielsen, G S Wilson
ABSTRACT
Low-level adsorption on the stationary phase has been studied using immunochemical reagents. An immunoaffinity column has been evaluated using affinity-purified radioisotope-labeled monoclonal antibodies. Recovery experiments including continuous immunosorbent monitoring have been performed. Proper characterization of an immunoaffinity separation can result in the recovery of immunologically active material in high yield.
Sigma-Aldrich
Iodine monochloride, 99.998% trace metals basis
Sigma-Aldrich
Iodine monochloride solution, 1 M in acetic acid
Sigma-Aldrich
Iodine monochloride, reagent grade, ≥95%
Sigma-Aldrich
Iodine monochloride solution, 1.0 M in methylene chloride
Sigma-Aldrich
Iodine monochloride, ACS reagent, 1.10±0.1 I/Cl ratio basis