Merck
  • Sustained recreational use of ecstasy is associated with altered pre and postsynaptic markers of serotonin transmission in neocortical areas: a PET study with [¹¹C]DASB and [¹¹C]MDL 100907.
Sustained recreational use of ecstasy is associated with altered pre and postsynaptic markers of serotonin transmission in neocortical areas: a PET study with [¹¹C]DASB and [¹¹C]MDL 100907.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2012-02-23)
Nina Bl Urban, Ragy R Girgis, Peter S Talbot, Lawrence S Kegeles, X Xu, W Gordon Frankle, Carl L Hart, Mark Slifstein, Anissa Abi-Dargham, Marc Laruelle
ABSTRACT
3,4-Methylenedioxymethamphetamine (MDMA), the main psychoactive component of the recreational drug ecstasy, is a potent serotonin (5-HT) releaser. In animals, MDMA induces 5-HT depletion and toxicity in 5-HT neurons. The aim of this study was to investigate both presynaptic (5-HT transporter, SERT) and postsynaptic (5-HT(2A) receptor) markers of 5-HT transmission in recently abstinent chronic MDMA users compared with matched healthy controls. We hypothesized that MDMA use is associated with lower SERT density and concomitant upregulation of 5-HT(2A) receptors. Positron emission tomography studies using the SERT ligand [¹¹C]DASB and the 5-HT(2A) receptor ligand [¹¹C]MDL 100907 were evaluated in 13 current and recently detoxified MDMA users and 13 matched healthy controls. MDMA users reported a mean duration of ecstasy use of 8 years, regular exposure, and at least 2 weeks of abstinence before the scans. SERT and 5-HT(2A) receptor availability (binding potential, BP(ND)) were analyzed with a two-tissue compartment model with arterial input function. Current recreational MDMA use was significantly associated with lower SERT BP(ND) and higher 5-HT(2A) receptor BP(ND) in cortical, but not subcortical regions. Decreased SERT BP(ND) was regionally associated with upregulated 5-HT(2A) receptor BP(ND). In light of the animal literature, the most parsimonious interpretation is that repeated exposure to MDMA in humans, even in moderate amounts, leads to damage in 5-HT neuron terminals innervating the cortex. Alterations in mood, cognition, and impulse control associated with these changes might contribute to sustain MDMA use. The reversibility of these changes upon abstinence remains to be firmly established.
Sigma-Aldrich
M100907, ≥98% (HPLC)