Merck
  • Fast single run of vanilla fingerprint markers on microfluidic-electrochemistry chip for confirmation of common frauds.
Fast single run of vanilla fingerprint markers on microfluidic-electrochemistry chip for confirmation of common frauds.
Electrophoresis (2009-09-10)
Mónica Avila, Mohammed Zougagh, Alberto Escarpa, Angel Ríos
ABSTRACT
A new strategy based on the fast separation of the fingerprint markers of Vanilla planifolia extracts and vanilla-related samples on microfluidic-electrochemistry chip is proposed. This methodology allowed the detection of all required markers for confirmation of common frauds in this field. The elution order was strategically connected with sequential sample screening and analyte confirmation steps, where first ethyl vanillin was detected to distinguish natural from adultered samples; second, vanillin as prominent marker in V. planifolia, but frequently added in its synthetic form; and third, the final detection of the fingerprint markers (p-hydroxybenzaldehyde, vanillic acid, and p-hydroxybenzoic acid) of V. planifolia with confirmation purposes. The reliability of the proposed methodology was demonstrated in the confirmation the natural or non-natural origin of vanilla in samples using V. planifolia extracts and other selected food samples containing this flavor.
Sigma-Aldrich
3-Ethoxy-4-hydroxybenzaldehyde, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl vanillin, ≥98%, FCC, FG