Merck
  • Cyanoacetic acid hydrazones of 3-(and 4-)acetylpyridine and some derived ring systems as potential antitumor and anti-HCV agents.
Cyanoacetic acid hydrazones of 3-(and 4-)acetylpyridine and some derived ring systems as potential antitumor and anti-HCV agents.
Archiv der Pharmazie (2006-01-18)
Soad A M El-Hawash, Abeer E Abdel Wahab, Maha A El-Demellawy
ABSTRACT
Two new acetylpyridinehydrazones derived from cyanoacetic acid hydrazide have been synthesized namely: cyanoacetic acid (1-pyridin-3 or 4-yl-ethylidene) hydrazides (1a,b). and some derived ring systems: 2-imino or 2-oxo-2H-chromenes (2a,b and 3a,b), substituted 2-thioxo-2,3-dihydrothiazoles (4a-d), substituted 2-thioxo-2,3-dihydro-6H-thiazolo[4,5-d]pyrimidin-7-ones (5a-d), substituted dihydrothiazoles (7a,b), and substituted 2-oxo-1,2-dihydropyridines (8a-d and 9a,b). Fifteen compounds were evaluated for their anticancer activity using the USA-NCI in-vitro screening program. Among the tested compounds, 8d exhibited a high value of percent tumor growth inhibition at concentrations of 10(-5) to 10(-7) M in all cancer cell lines, while 8b exhibited a significant value of percent tumor growth inhibition at concentration <10(-8 )M against non-small cells lung HOP-92. In addition, nine compounds were investigated for their in-vitro effect on the replication of hepatitis-C virus (HCV) in HepG2 hepatocellular carcinoma cell line infected with the virus using the reverse transcription polymerase chain reaction technique. Six compounds were capable of inhibiting the replication of both the HCV RNA (+)- and (-)-strands at 5-100 microg/mL concentration range. The activity order was 7b > 1b = 3a > 4c > 7a > 5c.
Sigma-Aldrich
Cyanoacetohydrazide, 98%
Sigma-Aldrich
3-Acetylpyridine, 98%
Sigma-Aldrich
3-Acetylpyridine, ≥98%, FG
Sigma-Aldrich
4-Acetylpyridine, 97%