Merck
  • High-intensity exercise training using a rotarod instrument (RotaHIIT) significantly improves exercise capacity in mice.
High-intensity exercise training using a rotarod instrument (RotaHIIT) significantly improves exercise capacity in mice.
Physiological reports (2024-05-03)
Jonathan J Herrera, Christopher M McAllister, Danielle Szczesniak, Rose-Carmel Goddard, Sharlene M Day
ABSTRACT
Voluntary or forced exercise training in mice is used to assess functional capacity as well as potential disease-modifying effects of exercise over a range of cardiovascular disease phenotypes. Compared to voluntary wheel running, forced exercise training enables precise control of exercise workload and volume, and results in superior changes in cardiovascular performance. However, the use of a shock grid with treadmill-based training is associated with stress and risk of injury, and declining compliance with longer periods of training time for many mouse strains. With these limitations in mind, we designed a novel, high-intensity interval training modality (HIIT) for mice that is carried out on a rotarod. Abbreviated as RotaHIIT, this protocol establishes interval workload intensities that are not time or resource intensive, maintains excellent training compliance over time, and results in improved exercise capacity independent of sex when measured by treadmill graded exercise testing (GXT) and rotarod specific acceleration and endurance testing. This protocol may therefore be useful and easily implemented for a broad range of research investigations. As RotaHIIT training was not associated cardiac structural or functional changes, or changes in oxidative capacity in cardiac or skeletal muscle tissue, further studies will be needed to define the physiological adaptations and molecular transducers that are driving the training effect of this exercise modality.
Sigma-Aldrich
Anti-GAPDH Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-TFAM (AB3) antibody produced in rabbit, affinity isolated antibody
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial