Merck
  • Interleukin-1RA Mitigates SARS-CoV-2-Induced Inflammatory Lung Vascular Leakage and Mortality in Humanized K18-hACE-2 Mice.
Interleukin-1RA Mitigates SARS-CoV-2-Induced Inflammatory Lung Vascular Leakage and Mortality in Humanized K18-hACE-2 Mice.
Arteriosclerosis, thrombosis, and vascular biology (2021-09-10)
Shiqin Xiong, Lianghui Zhang, Justin M Richner, Jake Class, Jalees Rehman, Asrar B Malik
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 7, ≥98%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
IL-1R Antagonist, The IL-1R Antagonist, also referenced under CAS 566914-00-9, controls the biological activity of IL-1R.