Merck
  • Reproducibility and accuracy of microscale thermophoresis in the NanoTemper Monolith: a multi laboratory benchmark study.
Reproducibility and accuracy of microscale thermophoresis in the NanoTemper Monolith: a multi laboratory benchmark study.
European biophysics journal : EBJ (2021-04-22)
Blanca López-Méndez, Bruno Baron, Chad A Brautigam, Thomas A Jowitt, Stefan H Knauer, Stephan Uebel, Mark A Williams, Arthur Sedivy, Olga Abian, Celeste Abreu, Malgorzata Adamczyk, Wojciech Bal, Sylvie Berger, Alexander K Buell, Carlo Carolis, Tina Daviter, Alexander Fish, Maria Garcia-Alai, Christian Guenther, Josef Hamacek, Jitka Holková, Josef Houser, Chris Johnson, Sharon Kelly, Andrew Leech, Caroline Mas, Daumantas Matulis, Stephen H McLaughlin, Roland Montserret, Rouba Nasreddine, Reine Nehmé, Quyen Nguyen, David Ortega-Alarcón, Kathryn Perez, Katja Pirc, Grzegorz Piszczek, Marjetka Podobnik, Natalia Rodrigo, Jasmina Rokov-Plavec, Susanne Schaefer, Tim Sharpe, June Southall, David Staunton, Pedro Tavares, Ondrej Vanek, Michael Weyand, Di Wu
ABSTRACT
Microscale thermophoresis (MST), and the closely related Temperature Related Intensity Change (TRIC), are synonyms for a recently developed measurement technique in the field of biophysics to quantify biomolecular interactions, using the (capillary-based) NanoTemper Monolith and (multiwell plate-based) Dianthus instruments. Although this technique has been extensively used within the scientific community due to its low sample consumption, ease of use, and ubiquitous applicability, MST/TRIC has not enjoyed the unambiguous acceptance from biophysicists afforded to other biophysical techniques like isothermal titration calorimetry (ITC) or surface plasmon resonance (SPR). This might be attributed to several facts, e.g., that various (not fully understood) effects are contributing to the signal, that the technique is licensed to only a single instrument developer, NanoTemper Technology, and that its reliability and reproducibility have never been tested independently and systematically. Thus, a working group of ARBRE-MOBIEU has set up a benchmark study on MST/TRIC to assess this technique as a method to characterize biomolecular interactions. Here we present the results of this study involving 32 scientific groups within Europe and two groups from the US, carrying out experiments on 40 Monolith instruments, employing a standard operation procedure and centrally prepared samples. A protein-small molecule interaction, a newly developed protein-protein interaction system and a pure dye were used as test systems. We characterized the instrument properties and evaluated instrument performance, reproducibility, the effect of different analysis tools, the influence of the experimenter during data analysis, and thus the overall reliability of this method.
Product Description
Roche
Lysozyme, from hen egg white
Pricing and availability is not currently available.