Merck
  • Formin Activity and mDia1 Contribute to Maintain Axon Initial Segment Composition and Structure.
Formin Activity and mDia1 Contribute to Maintain Axon Initial Segment Composition and Structure.
Molecular neurobiology (2021-08-31)
Wei Zhang, María Ciorraga, Pablo Mendez, Diana Retana, Norah Boumedine-Guignon, Beatriz Achón, Michaël Russier, Dominique Debanne, Juan José Garrido
ABSTRACT
The axon initial segment (AIS) is essential for maintaining neuronal polarity, modulating protein transport into the axon, and action potential generation. These functions are supported by a distinctive actin and microtubule cytoskeleton that controls axonal trafficking and maintains a high density of voltage-gated ion channels linked by scaffold proteins to the AIS cytoskeleton. However, our knowledge of the mechanisms and proteins involved in AIS cytoskeleton regulation to maintain or modulate AIS structure is limited. In this context, formins play a significant role in the modulation of actin and microtubules. We show that pharmacological inhibition of formins modifies AIS actin and microtubule characteristics in cultured hippocampal neurons, reducing F-actin density and decreasing microtubule acetylation. Moreover, formin inhibition diminishes sodium channels, ankyrinG and βIV-spectrin AIS density, and AIS length, in cultured neurons and brain slices, accompanied by decreased neuronal excitability. We show that genetic downregulation of the mDia1 formin by interference RNAs also decreases AIS protein density and shortens AIS length. The ankyrinG decrease and AIS shortening observed in pharmacologically inhibited neurons and neuron-expressing mDia1 shRNAs were impaired by HDAC6 downregulation or EB1-GFP expression, known to increase microtubule acetylation or stability. However, actin stabilization only partially prevented AIS shortening without affecting AIS protein density loss. These results suggest that mDia1 maintain AIS composition and length contributing to the stability of AIS microtubules.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Synaptopodin (SE-19) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Tubastatin A hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
SMIFH2, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-Sodium Channel, Pan antibody produced in mouse, ~1 mg/mL, clone K58/35, purified immunoglobulin
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
bisBenzimide H 33342 trihydrochloride, ≥98% (HPLC and TLC)