Merck
  • Study on the substituents' effects of a series of synthetic chalcones against the yeast Candida albicans.
Study on the substituents' effects of a series of synthetic chalcones against the yeast Candida albicans.
European journal of medicinal chemistry (2006-09-30)
D Batovska, St Parushev, A Slavova, V Bankova, I Tsvetkova, M Ninova, H Najdenski
ABSTRACT
A large series of chalcones were synthesized and studied for activity against Candida albicans. The SAR analysis showed that the antifungal activity was highly dependent on the substitution pattern of the aryl rings and correlated to a large extent with the ability of compounds to interact with sulfhydryl groups. The most active were the hydroxylated chalcones as their activity related to the location of the phenolic group in the aryl ring B as follows: o-OH>p-OH approximately 3,4-di-OH>m-OH. These and other correlations obtained strongly contribute to the knowledge for design of anticandidal chalcones.
Sigma-Aldrich
trans-Chalcone, 97%
Sigma-Aldrich
trans,trans-Dibenzylideneacetone, 98%