Merck
Selection of novel structural zinc sites from a random peptide library.
FEBS letters (2003-12-04)
Teruhiko Matsubara, Yuko Hiura, Osamu Kawahito, Mikito Yasuzawa, Katsuhiro Kawashiro
ABSTRACT
Zinc ion (Zn(2+)) can be coordinated with four or three amino acid residues to stabilize a protein's structure or to form a catalytic active center. We used phage display selection of a dodecamer random peptide library with Zn(2+) to identify structural zinc sites. The binding specificity for Zn(2+) of selected sequences was confirmed using enzyme-linked immunosorbent and competitive inhibition assays. Circular dichroism spectra indicated that the interaction with Zn(2+) induced a change in conformation, which means the peptide acts as a structural zinc site. Furthermore, a search of protein databases revealed that two selected sequences corresponded to parts of natural zinc sites of copper/zinc superoxide dismutase and zinc-containing ferredoxin. We demonstrated that Zn(2+)-binding sequences selected from the random combinatorial library would be candidates for artificial structural zinc sites.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-fd Bacteriophage antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution