Merck
  • Reduced IgE and IgG antigenic response to milk proteins hydrolysates obtained with the use of non-commercial serine protease from Yarrowia lipolytica.
Reduced IgE and IgG antigenic response to milk proteins hydrolysates obtained with the use of non-commercial serine protease from Yarrowia lipolytica.
Food chemistry (2019-08-16)
Anna Dąbrowska, Joanna Bajzert, Konrad Babij, Marek Szołtysik, Tadeusz Stefaniak, Ewa Willak-Janc, Józefa Chrzanowska
ABSTRACT
The aim of the study was to investigate the use of serine protease from Yarrowia lipolytica yeast for reduction of milk proteins allergenicity. Whey protein concentrate (WPC-80), αs-casein and their hydrolysates were analyzed for the capacity to bind IgE and IgG antibodies present in sera from patients with cow milk protein allergy using a competitive ELISA. The hydrolysis of αs-casein and whey protein concentrate contributed to a significant reduction of their immunoreactive epitopes. In case of IgE antibodies, the lowest binding capacity was detected in the 24 h hydrolysates of both proteins in which the inhibition of the reaction was ≤20 and ≤68% for αs-casein and whey protein concentrate respectively. One hour hydrolysis of WPC-80 reduced the protein antigenicity, while the longer time (5 h) might lead to the exposure of new IgE - reactive epitopes.
Sigma-Aldrich
3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA, ready to use solution
Sigma-Aldrich
Protein standard, liquid, 2 mg protein/ml