Merck
  • Influence of deep frying on the unsaponifiable fraction of vegetable edible oils enriched with natural antioxidants.
Influence of deep frying on the unsaponifiable fraction of vegetable edible oils enriched with natural antioxidants.
Journal of agricultural and food chemistry (2011-06-08)
Mara I Orozco, Feliciano Priego-Capote, Maria D Luque de Castro
ABSTRACT
The influence of deep frying, mimicked by 20 heating cycles at 180 °C (each cycle from ambient temperature to 180 °C maintained for 5 min), on the unsaponifiable fraction of vegetable edible oils represented by three characteristic families of compounds (namely, phytosterols, aliphatic alcohols, and triterpenic compounds) has been studied. The target oils were extra virgin olive oil (with intrinsic content of phenolic antioxidants), refined sunflower oil enriched with antioxidant phenolic compounds isolated from olive pomace, refined sunflower oil enriched with an autoxidation inhibitor (dimethylpolysiloxane), and refined sunflower oil without enrichment. Monitoring of the target analytes as a function of both heating cycle and the presence of natural antioxidants was also evaluated by comparison of the profiles after each heating cycle. Identification and quantitation of the target compounds were performed by gas cromatography-mass spectrometry in single ion monitoring mode. Analysis of the heated oils revealed that the addition of natural antioxidants could be an excellent strategy to decrease degradation of lipidic components of the unsaponifiable fraction with the consequent improvement of stability.
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 500 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 50 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 200 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 1,000 cSt (25 °C)(lit.)
Sigma-Aldrich
Dimethylpolysiloxane, viscosity 5 cSt (25 °C)(lit.)