Skip to Content
Merck
vibrant-m

TSKgel® BioAssist® Chelate (10 µm) HPLC Columns

vibrant-m
Synonym(s):
TSKgel® Chelate-5PW HPLC Column and Guardgel Kit

Products

Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing