
Sodium aminosalicylate dihydrate

Synonym(s):
Sodium 4-aminosalicylate dihydrate, 4-Amino-2-hydroxybenzoic acid sodium salt, 4-Amino-salicylic acid sodium salt
Empirical Formula (Hill Notation):
C7H6NNaO3 · 2H2O
CAS Number:
Molecular Weight:
211.15
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing