
Simethicone

Synonym(s):
α-(Trimethylsilyl)-ω-methylpoly[oxy(dimethylsilylene)] mixture with silicon dioxide, Antifoam A, Antifoam AF
CAS Number:
Products
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing