
S-Acetylthioglycolic acid N-hydroxysuccinimide ester

Synonym(s):
N-Succinimidyl (acetylthio)acetate, N-Succinimidyl S-acetylthioglycolate, S-Acetylthioglycolic acid NHS ester, SATA
Empirical Formula (Hill Notation):
C8H9NO5S
CAS Number:
Molecular Weight:
231.23
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing