
Rifabutin impurity A

Synonym(s):
1-(2-Methylpropyl)-4-piperidone
Empirical Formula (Hill Notation):
C9H17NO
CAS Number:
Molecular Weight:
155.24
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing