
Rhenium

Synonym(s):
Rhenium
Empirical Formula (Hill Notation):
Re
CAS Number:
Molecular Weight:
186.21
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Quality Segment
Showing 1-20 of 81
Page 1 of 5
Compare
Product Number
Description
Pricing
Page 1 of 5