R-Citalopram oxalate
Synonym(s):
(R)-Citalopram oxalate salt, (R)-1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile
Empirical Formula (Hill Notation):
C20H21FN2O Ā· C2H2O4
CAS Number:
Molecular Weight:
414.43
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing