
Racecadotril

Synonym(s):
(RS)-Benzyl N-[3-(acetylthio)-2-benzylpropanoyl]glycinate, (±)-Acetorphan, Acetorphan, Cadotril, Redotil, Tiorfan, Zedott
Empirical Formula (Hill Notation):
C21H23NO4S
CAS Number:
Molecular Weight:
385.48
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Format
Research Area
Application
API Family
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing