
Quinapril for peak identification

Synonym(s):
Quinapril hydrochloride, (3S)-2-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)-3-phenylpropyl]amino]-1-oxopropyl]-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C25H30N2O5 · HCl
CAS Number:
Molecular Weight:
474.98
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing