![Phenazone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/109/411/e37c0127-8cfa-42b4-be35-a19e920314b1/640/e37c0127-8cfa-42b4-be35-a19e920314b1.png)
Phenazone
![Phenazone European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/109/411/e37c0127-8cfa-42b4-be35-a19e920314b1/640/e37c0127-8cfa-42b4-be35-a19e920314b1.png)
Synonym(s):
Antipyrine, 2,3-Dimethyl-1-phenyl-3-pyrazolin-5-one, Phenazone
Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing