Mevastatin/Lovastatin Impurity A
Synonym(s):
Mevastatin, Compactin, ML-236B
Empirical Formula (Hill Notation):
C23H34O5
CAS Number:
Molecular Weight:
390.51
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing