Skip to Content
Merck
Meperidine-D4 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Meperidine-D4 solution

Meperidine-D4 solution 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C15D4H17NO2
CAS Number:
Molecular Weight:
251.36
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique