
Isoamyl hexanoate, mixture of isomers

Empirical Formula (Hill Notation):
C11H22O2
CAS Number:
Molecular Weight:
186.29
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing