![Hydrocodone-D6 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/289/258/6449126d-f70b-4b79-85e2-2283ed72e1a2/640/6449126d-f70b-4b79-85e2-2283ed72e1a2.png)
Hydrocodone-D6 solution
![Hydrocodone-D6 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/289/258/6449126d-f70b-4b79-85e2-2283ed72e1a2/640/6449126d-f70b-4b79-85e2-2283ed72e1a2.png)
Empirical Formula (Hill Notation):
C18H15NO3D6
CAS Number:
Molecular Weight:
305.40
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2