Skip to Content
Merck
Guanosine 5′-diphospho-β-L-fucose sodium salt ≥85%

Guanosine 5′-diphospho-β-L-fucose sodium salt

Guanosine 5′-diphospho-β-L-fucose sodium salt ≥85%
Synonym(s):
6-Deoxy-β-L-galactopyranosylguanosine 5′-diphosphate, GDP-Fuc, GDP-fucose
Empirical Formula (Hill Notation):
C16H23N5Na2O15P2
CAS Number:
Molecular Weight:
633.31

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing