
Dulbecco’s Modified Eagle’s Medium - low glucose

Synonym(s):
DME, DMEM
Products
Product Category
Brand
Physical Form
Glucose (g/L)
HEPES (mM)
L-Glutamine
Sodium Bicarbonate
Phenol Red
Sodium Pyruvate (g/L)
Quality Segment
Sterilization Method
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing