Skip to Content
Merck
(±)-SKF-38393 hydrochloride crystalline, ≥98% (HPLC)

(±)-SKF-38393 hydrochloride

(±)-SKF-38393 hydrochloride crystalline, ≥98% (HPLC)
Synonym(s):
(±)-1-Phenyl-2,3,4,5-tetrahydro-(1H)-3-benzazepine-7,8-diol hydrochloride
Empirical Formula (Hill Notation):
C16H17NO2 · HCl
CAS Number:
Molecular Weight:
291.77

Products

Product Category
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing