
Diphenhydramine Related Compound B

Synonym(s):
(RS)-3-(Diphenylmethoxy)-1,2-propanediol, 3-(Benzhydryloxy)propane-1,2-diol
Empirical Formula (Hill Notation):
C16H18O3
CAS Number:
Molecular Weight:
258.31
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2