
Diphenhydramine N-Oxide Hydrochloride

Synonym(s):
Diphenhydramine N-oxide hydrochloride, 2-(Diphenylmethoxy)-N,N-dimethylethylamine N-oxide hydrochloride
Empirical Formula (Hill Notation):
C17H21NO2 · HCl
CAS Number:
Molecular Weight:
307.82
Products
Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing