![Diphenhydramine N-oxide United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/392/490/8e4a9c4c-9505-422d-8be4-ebb1af127258/640/8e4a9c4c-9505-422d-8be4-ebb1af127258.png)
Diphenhydramine N-oxide
![Diphenhydramine N-oxide United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/392/490/8e4a9c4c-9505-422d-8be4-ebb1af127258/640/8e4a9c4c-9505-422d-8be4-ebb1af127258.png)
Synonym(s):
Diphenhydramine N-oxide hydrochloride, 2-(Diphenylmethoxy)-N,N-dimethylethylamine N-oxide hydrochloride
Empirical Formula (Hill Notation):
C17H21NO2 · HCl
CAS Number:
Molecular Weight:
307.82
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing