
Dibutyl disulfide

Synonym(s):
Butyl disulfide, Dibutyl disulfide
Linear Formula:
CH3(CH2)3SS(CH2)3CH3
CAS Number:
Molecular Weight:
178.36
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing