
Coumestrol

Synonym(s):
7,12-Dihydroxycoumestan
Empirical Formula (Hill Notation):
C15H8O5
CAS Number:
Molecular Weight:
268.22
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing