
Butyl stearate

Synonym(s):
Stearic acid butyl ester
Linear Formula:
CH3(CH2)16COO(CH2)3CH3
CAS Number:
Molecular Weight:
340.58
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing