
Boc-Tyr-OH

Synonym(s):
N-(tert-Butoxycarbonyl)-L-tyrosine, Boc-L-tyrosine, Boc-Tyr-OH
Linear Formula:
4-(HO)C6H4CH2CH(COOH)NHC(O)OC(CH3)3
CAS Number:
Molecular Weight:
281.30
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing