
Baclofen impurity A

Synonym(s):
(4RS)-4-(4-Chlorophenyl)pyrrolidin-2-one, 4-(4-Chlorophenyl)-2-pyrrolidinone
Empirical Formula (Hill Notation):
C10H10ClNO
CAS Number:
Molecular Weight:
195.65
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing