
Atrazine-2-hydroxy

Synonym(s):
2-Hydroxyatrazine
Empirical Formula (Hill Notation):
C8H15N5O
CAS Number:
Molecular Weight:
197.24
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing