![Albendazole-2-aminosulfone ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/382/949/6f4b6003-4459-485e-9427-bb0c5124e375/640/6f4b6003-4459-485e-9427-bb0c5124e375.png)
Albendazole-2-aminosulfone
![Albendazole-2-aminosulfone ≥95% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/382/949/6f4b6003-4459-485e-9427-bb0c5124e375/640/6f4b6003-4459-485e-9427-bb0c5124e375.png)
Synonym(s):
2-Amino-5-propylsulfonylbenzimidazole, 2-Aminoalbendazole sulfone
Empirical Formula (Hill Notation):
C10H13N3O2S
CAS Number:
Molecular Weight:
239.29
Products
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing