
α-Humulene

Synonym(s):
α-Caryophyllene, trans,trans,trans-2,6,6,9-Tetramethyl-1,4,8-cycloundecatriene
Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing