
4,6-Dioxoheptanoic acid

Synonym(s):
Succinylacetone
Linear Formula:
CH3COCH2COCH2CH2CO2H
CAS Number:
Molecular Weight:
158.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing