
3-Methoxy-L-tyrosine monohydrate

Synonym(s):
4-Hydroxy-3-methoxy-L-phenylalanine, L-3-O-Methyl-DOPA
Empirical Formula (Hill Notation):
C10H13NO4 H2O
CAS Number:
Molecular Weight:
229.23
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-1 of 1
Compare
Product Number
Description
Pricing